Molybdenum 99,95 Metals basis, plate, Thickness: 2.5 mm (0.098 in)
Supplier: Thermo Scientific Chemicals
10047-FI
10047-GH
10047-HP
CAAA10047-FIEA
237
CAD
CAAA10047-FI
CAAA10047-GH
CAAA10047-HP
Molybdenum 99,95 Metals basis, plate, Thickness: 2.5 mm (0.098 in)
Molybdenum
MDL: MFCD00003465
Purity: 99.95%
CAS No.: 7439-98-7
Physical Form: ≈64g/50x50mm
MDL No.: MFCD00003465
Formula:
Mo MW: 95.94 g/mol Boiling Pt: 3617 Melting Pt: 2622 Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...