Molybdenum ≥99.95% (metals basis), foil, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

10046-FI 10046-GH 10046-GW
CAAA10046-FIEA 117.39 CAD
CAAA10046-FI CAAA10046-GH CAAA10046-GW
Molybdenum ≥99.95% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈25.6g/50x50mm MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR