Molybdenum ≥99.95% (metals basis), foil, annealed, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10044-GH 10044-HW 10044-HX
CAAA10044-GHEA 120.4 CAD
CAAA10044-GH CAAA10044-HW CAAA10044-HX
Molybdenum ≥99.95% (metals basis), foil, annealed, Thickness: 0.25 mm (0.01 in)
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈25.6g/100x100mm, Temper: soft MDL No.: MFCD00003465

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR