Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

10043-HW 10043-HX
CAAA10043-HWEA 243.32 CAD
CAAA10043-HW CAAA10043-HX
Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈10.2g/100x100mm MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR