Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

10042-GT 10042-HB 10042-HZ
CAAA10042-GTEA 156.1 CAD
CAAA10042-GT CAAA10042-HB CAAA10042-HZ
Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈5.8g/150x150mm MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR