Molybdenum 99,94 Metals basis, wire, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
10039-G5
10039-G9
10039-H4
CAAA10039-G5EA
134.14
CAD
CAAA10039-G5
CAAA10039-G9
CAAA10039-H4
Molybdenum 99,94 Metals basis, wire, annealed, Ø 1.0 mm (0.04 in)
Molybdenum
MDL: MFCD00003465
Purity: 99.94%
CAS No.: 7439-98-7
Physical Form: ≈8.0g/m, Temper: soft
MDL No.: MFCD00003465
Formula:
Mo MW: 95.94 g/mol Boiling Pt: 3617 Melting Pt: 2622 Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...