Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
10038-G9
10038-H4
CAAA10038-G9EA
178.54
CAD
CAAA10038-G9
CAAA10038-H4
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Molybdenum
MDL: MFCD00003465
Purity: 99.95%
CAS No.: 7439-98-7
Physical Form: ≈2.0g/m, Temper: soft
MDL No.: MFCD00003465
Formula:
Mo MW: 95.94 g/mol Boiling Pt: 3617 Melting Pt: 2622 Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...