Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10036-G9 10036-H4
CAAA10036-G9EA 114.55 CAD
CAAA10036-G9 CAAA10036-H4
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈0.50g/m, Temper: soft MDL No.: MFCD00003465

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR