Molybdenum ≥99.95% (metals basis), powder APS 3-7 µm

Supplier: Thermo Scientific Chemicals
Warning

10030-06 10030-18 10030-30 10030-A1
CAAA10030-06EA 51.03 CAD
CAAA10030-06 CAAA10030-18 CAAA10030-30 CAAA10030-A1
Molybdenum ≥99.95% (metals basis), powder APS 3-7 µm
Molybdenum

MDL: MFCD00003465


Purity: 99.95% CAS No.: 7439-98-7 Appearance: Grey Odor: Odorless UN No.: 3089 MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 5560 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR