Cesium sulfate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
10027-14
10027-22
CAAA10027-14EA
118.97
CAD
CAAA10027-14
CAAA10027-22
Cesium sulfate ≥99.9% (metals basis)
Cesium sulphate
MDL: MFCD00010959
Very soluble in water. Slightly soluble in alcohol, acetone, pyridine.
Purity: 99.9%
CAS No.: 10294-54-9
Molecular Formula: Cs2SO4
Formula Weight: 361.87
Physical Form: -20 Mesh Powder
Density: 4.243
Melting Point: 1010°
Solubility: Very soluble in water. Slightly soluble in alcohol, acetone, pyridine
Merck Reference: 14,2016
MDL No.: MFCD00010959
Formula:
Cs₂O₄S MW: 361.87 g/mol Boiling Pt: 1627 Melting Pt: 1010 Density: 4.243 Storage Temperature: Ambient |
MDL Number:
MFCD00010959 CAS Number: 10294-54-9 EINECS: 233-662-6 Merck Index: 14,2016 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...