Cesium iodide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00010962
Very soluble in water. Soluble in ethanol. Slightly soluble in methanol. Insoluble in acetone
Purity: 99.9%
CAS No.: 7789-17-5
Molecular Formula: CsI
Formula Weight: 259.81
Physical Form: -20 Mesh
Refractive Index: 1.7876
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 626°
Boiling Point: 1280°
Solubility: Very soluble in water. Soluble in ethanol. Slightly soluble in methanol. Insoluble in acetone
Merck Reference: 14,2014
MDL No.: MFCD00010962
Formula:
CsI Boiling Pt: 1280 °C Melting Pt: 626 °C Density: 3.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 EINECS: 232-145-2 Merck Index: 14,2014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...