Cesium hydroxide 50% w/w aqueous solution , 99.9% Metals basis

Supplier: Thermo Scientific Chemicals
Danger

10020-ND 10020-NF
CAAA10020-NDEA 273.34 CAD
CAAA10020-ND CAAA10020-NF
Cesium hydroxide 50% w/w aqueous solution , 99.9% Metals basis
Cesium hydroxide

MDL: MFCD00010964 Notes: Sold on Cesium hydroxide contained basis


Purity: 99.9% CAS No.: 21351-79-1 Molecular Formula: CsOH Formula Weight: 149.91 Physical Form: Liquid Density: 1.725 Appearance: Colorless Odor: Odorless UN No.: 2681 MDL No.: MFCD00010964

Formula: CsOH
MW: 149.91
Melting Pt: 338.85
Density: 1.725
Storage Temperature: Ambient
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR