Beryllium oxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10001-06
10001-14
CAAA10001-06EA
80.58
CAD
CAAA10001-06
CAAA10001-14
Beryllium oxide ≥99% (metals basis)
Beryllium oxide
MDL: MFCD00003457
Insoluble in water. Slowly soluble in concentrated acids, alkali hydroxides
Purity: 99%
CAS No.: 1304-56-9
Molecular Formula: BeO
Formula Weight: 25.01
Physical Form: Powder
Density: 3.01
Melting Point: 2575°
Boiling Point: 4300°
Solubility: Insoluble in water. Slowly soluble in concentrated acids, alkali hydroxides
Merck Reference: 14,1172
UN No.: 1566
MDL No.: MFCD00003457
Formula:
BeO Boiling Pt: 4300 °C Melting Pt: 2575 °C Density: 3.01 Storage Temperature: Ambient |
MDL Number:
MFCD00003457 CAS Number: 1304-56-9 EINECS: 215-133-1 UN: 1566 ADR: 6.1,II Merck Index: 14,1172 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...