Tantalum ≥99.9% (metals basis), wire, Ø 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00977-G1 00977-G5
CAAA00977-G1EA 107.6 CAD
CAAA00977-G1 CAAA00977-G5
Tantalum ≥99.9% (metals basis), wire, Ø 0.127 mm (0.005 in)
Tantalum

MDL: MFCD00011252


Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈0.21g/m Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR