Titanium ≥99% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals
Warning

00971-FI 00971-GT
CAAA00971-FIEA 76.47 CAD
CAAA00971-FI CAAA00971-GT
Titanium ≥99% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Titanium

MDL: MFCD00011264


Purity: 99% CAS No.: 7440-32-6 Physical Form: ≈1.4g/50x50mm Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR