Tin ≥99.85% (metals basis), powder -100 mesh

Supplier: Thermo Scientific Chemicals

00941-22 00941-36
CAAA00941-22EA 100.49 CAD
CAAA00941-22 CAAA00941-36
Tin ≥99.85% (metals basis), powder -100 mesh
Tin

MDL: MFCD00133862


Purity: 99.85% CAS No.: 7440-31-5 Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR