Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

00939-G2 00939-G6
CAAA00939-G2EA 79.63 CAD
CAAA00939-G2 CAAA00939-G6
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 1.0 mm (0.04 in)
Zirconium

MDL: MFCD00011303


Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈5.11g/m Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR