Zirconium ≥99.2% (metals basis excluding Hf), plate 25x100 mm (0.98x3.9 in), thickness 6.4 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
00934-KF
00934-KJ
CAAA00934-KFEA
194.34
CAD
CAAA00934-KF
CAAA00934-KJ
Zirconium ≥99.2% (metals basis excluding Hf), plate 25x100 mm (0.98x3.9 in), thickness 6.4 mm (0.25 in)
Zirconium
MDL: MFCD00011303
Purity: 99.2%
CAS No.: 7440-67-7
Physical Form: ≈104g/25x100mm, Temper: annealed
Odor: Odorless
MDL No.: MFCD00011303
Formula:
Zr Storage Temperature: Ambient |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...