Tantalum ≥99.98% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
00933-KF
00933-KJ
CAAA00933-KFEA
71.1
CAD
CAAA00933-KF
CAAA00933-KJ
Tantalum ≥99.98% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Tantalum
MDL: MFCD00011252
Purity: 99.98%
CAS No.: 7440-25-7
Physical Form: ≈6.8g/slug
Odor: Odorless
MDL No.: MFCD00011252
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...