Tin ≥99.8% (metals basis), ingot 2.54x0.95x30.5 cm (1.0x0.37x12.0 in)

Supplier: Thermo Scientific Chemicals

00929-KF 00929-KJ
CAAA00929-KFEA 149.63 CAD
CAAA00929-KF CAAA00929-KJ
Tin ≥99.8% (metals basis), ingot 2.54x0.95x30.5 cm (1.0x0.37x12.0 in)
Tin

MDL: MFCD00133862


Purity: 99.8% CAS No.: 7440-31-5 Physical Form: Cast ingot, dimensions are approximate Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR