Bismuth ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
00915-30
00915-A1
CAAA00915-30EA
303.36
CAD
CAAA00915-30
CAAA00915-A1
Bismuth ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Bismuth
MDL: MFCD00134033
Purity: 99.9%
CAS No.: 7440-69-9
Physical Form: ≈12.5g/cm
Odor: Odorless
MDL No.: MFCD00134033
Formula:
Bi Boiling Pt: 1490 °C (1013 hPa) Melting Pt: 271…272 °C Density: 9.78 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 Merck Index: 12,01297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...