Bromine ≥99.8%, liquid

Supplier: Thermo Scientific Chemicals

00905-A1 00905-30 00905-14 00905-30 00905-14 00905-A1 00905-D9 00905-D9
CAAA00905-30EA 122.92 CAD
CAAA00905-30 AA00905-A1 AA00905-30 CAAA00905-A1 AA00905-14 CAAA00905-D9 CAAA00905-14 AA00905-D9
Bromine ≥99.8%, liquid
Bromine
Formula: Br₂
MW: 159.81 g/mol
Boiling Pt: 58.7 °C (1013 hPa)
Melting Pt: –7.2 °C
Density: 3.12 g/cm³ (20 °C)
MDL Number: MFCD00010896
CAS Number: 7726-95-6
UN: 1744
ADR: 8,I
Merck Index: 13,01378

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR