Magnesium ≥99.8% (metals basis), granules -12+50 mesh

Supplier: Thermo Scientific Chemicals
Danger

00870-36 00870-A1 00870-A3
CAAA00870-36EA 200.66 CAD
CAAA00870-36 CAAA00870-A1 CAAA00870-A3
Magnesium ≥99.8% (metals basis), granules -12+50 mesh
Magnesium

MDL: MFCD00085308


Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver-grey Odor: Odorless UN No.: 1869 MDL No.: MFCD00085308

Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR