Gold ≥99.95% (metals basis), sponge -20 mesh
Supplier: Thermo Scientific Chemicals
00827-03
00827-06
CAAA00827-03EA
453.46
CAD
CAAA00827-03
CAAA00827-06
Gold ≥99.95% (metals basis), sponge -20 mesh
Gold
MDL: MFCD00003436
Purity: 99.95%
CAS No.: 7440-57-5
MDL No.: MFCD00003436
Formula:
Au Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...