Silver ≥99.9% (metals basis), soluble APS 4-8 µm

Supplier: Thermo Scientific Chemicals

00783-09 00783-18
CAAA00783-09EA 111.86 CAD
CAAA00783-09 CAAA00783-18
Silver ≥99.9% (metals basis), soluble APS 4-8 µm
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Odor: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR