Silver ≥99.999% (metals basis), powder, Premion® -120 mesh

Supplier: Thermo Scientific Chemicals

00781-06 00781-06 00781-14 00781-14
CAAA00781-14EA 557.74 CAD
AA00781-14 CAAA00781-14 AA00781-06 CAAA00781-06
Silver ≥99.999% (metals basis), powder, Premion® -120 mesh
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961.9 °C
Density: 10.5 g/cm³ (20 °C)
MDL Number: MFCD00003397
CAS Number: 7440-22-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR