Lead ≥99.999% (metals basis), rods, Ø 9.5 mm (0.37 in)

Supplier: Thermo Scientific Chemicals
Danger

00770-2Z 00770-DW
CAAA00770-2ZEA 178.54 CAD
CAAA00770-2Z CAAA00770-DW
Lead ≥99.999% (metals basis), rods, Ø 9.5 mm (0.37 in)
Lead

MDL: MFCD00134050


Purity: 99.999+% CAS No.: 7439-92-1 Physical Form: ≈7.8g/cm Appearance: Grey Odor: Odorless MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR