Gold ≥99.95% (metals basis), spherical powder <5 µm

Supplier: Thermo Scientific Chemicals

00765-03 00765-06
CAAA00765-03EA 453.46 CAD
CAAA00765-03 CAAA00765-06
Gold ≥99.95% (metals basis), spherical powder <5 µm
Gold

MDL: MFCD00003436


Purity: 99.95% CAS No.: 7440-57-5 Physical Form: S.A. 0.10-0.35m2/g MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR