Lithium 99% (metals basis), granules 1-6 mm (0.04-0.2 in)

Supplier: Thermo Scientific Chemicals
Danger

00733-18 00733-30
CAAA00733-18EA 213.52 CAD
CAAA00733-18 CAAA00733-30
Lithium 99% (metals basis), granules 1-6 mm (0.04-0.2 in)
Lithium

MDL: MFCD00134051 Notes: Sodium is major impurity


Purity: 99% CAS No.: 7439-93-2 Physical Form: Packaged under argon Appearance: Silvery Odor: Odorless UN No.: 1415 MDL No.: MFCD00134051

Formula: Li
MW: 6.94 g/mol
Boiling Pt: 1317
Melting Pt: 179
Density: 0.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134051
CAS Number: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3 I
Merck Index: 13,05542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR