Titanium ≥93%, powder <20 µm
Supplier: Thermo Scientific Chemicals
|
Danger
|
00681-14
00681-22
00681-36
CAAA00681-14EA
229.1
CAD
CAAA00681-14
CAAA00681-22
CAAA00681-36
Titanium ≥93%, powder <20 µm
Titanium
Purity: 93%
CAS No.: 7440-32-6
Physical Form: Packaged in water
Odor: Odorless
UN No.: 1352
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3277 °C (1013 hPa) Melting Pt: 1677 °C Density: 4.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 1352 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...