Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)

Supplier: Thermo Scientific Chemicals
Danger

00653-06 00653-18 00653-30
CAAA00653-06EA 65.89 CAD
CAAA00653-06 CAAA00653-18 CAAA00653-30
Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)
Barium

MDL: MFCD00134031


Purity: 99.2% CAS No.: 7440-39-3 Physical Form: Packaged in oil Appearance: Yellow-silver Odor: Odorless UN No.: 1400 MDL No.: MFCD00134031

Formula: Ba
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR