Lutetium ≥99.9% (REO, rare earth oxide basis), pieces

Supplier: Thermo Scientific Chemicals

00628-03 00628-06
CAAA00628-03EA 366.56 CAD
CAAA00628-03 CAAA00628-06
Lutetium ≥99.9% (REO, rare earth oxide basis), pieces
Lutetium

MDL: MFCD00011098


Purity: 99.9% CAS No.: 7439-94-3 Physical Form: Packaged in air-stable glass bottle Odor: Odorless MDL No.: MFCD00011098

Formula: Lu
MW: 174.97 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1651…1675 °C
Density: 9.849 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR