Copper ≥99% (metals basis), powder -625 mesh (APS 2-3,50 µm)
Supplier: Thermo Scientific Chemicals
|
Warning
|
00622-09
00622-18
00622-30
CAAA00622-09EA
53.09
CAD
CAAA00622-09
CAAA00622-18
CAAA00622-30
Copper ≥99% (metals basis), powder -625 mesh (APS 2-3,50 µm)
Copper
MDL: MFCD00010965
Purity: 99%
CAS No.: 7440-50-8
Physical Form: Packaged under argon
Appearance: Reddish
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 UN: 3089 ADR: 4.1 III Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...