Yttrium ≥99.9% (REO, rare earth oxide basis), pieces
Supplier: Thermo Scientific Chemicals
00615-09
00615-18
CAAA00615-09EA
233.84
CAD
CAAA00615-09
CAAA00615-18
Yttrium ≥99.9% (REO, rare earth oxide basis), pieces
Yttrium
MDL: MFCD00011468
Purity: 99.9%
CAS No.: 7440-65-5
Physical Form: Packaged under argon
Odor: Odorless
MDL No.: MFCD00011468
Formula:
Y Boiling Pt: 2927 °C (1013 hPa) Melting Pt: 1525 °C Density: 4.472 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 Merck Index: 12,10240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...