Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Supplier: Thermo Scientific Chemicals
00610-FF
00610-FI
CAAA00610-FFEA
510.34
CAD
CAAA00610-FF
CAAA00610-FI
Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Samarium
MDL: MFCD00011233
Purity: 99.9%
CAS No.: 7440-19-9
Physical Form: ≈2.9g/25x25mm, Packaged in mylar under argon
Odor: Odorless
MDL No.: MFCD00011233
Formula:
Sm Boiling Pt: 1900 °C (1013 hPa) Melting Pt: 1076.85 °C Density: 7.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...