Samarium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 12 mm (0.47 in)
Supplier: Thermo Scientific Chemicals
00609-09
00609-18
CAAA00609-03EA
53.18
CAD
CAAA00609-03
CAAA00609-09
CAAA00609-18
Samarium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 12 mm (0.47 in)
Samarium
MDL: MFCD00011233
Purity: 99.9%
CAS No.: 7440-19-9
Physical Form: Packaged in air stable poly bottle
Sensitivity: Air and Moisture Sensitive
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00011233
Formula:
Sm Boiling Pt: 1900 °C (1013 hPa) Melting Pt: 1076.85 °C Density: 7.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...