Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.62 mm (0.024 in)
Supplier: Thermo Scientific Chemicals
00595-FF
00595-FI
CAAA00595-FFEA
293.88
CAD
CAAA00595-FF
CAAA00595-FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.62 mm (0.024 in)
Gadolinium
MDL: MFCD00011022
Purity: 99.9%
CAS No.: 7440-54-2
Physical Form: ≈3.1g/25x25mm, Packaged under argon
MDL No.: MFCD00011022
Formula:
Gd MW: 157.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...