Erbium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.62 mm (0.024 in)
Supplier: Thermo Scientific Chemicals
00593-FF
00593-FI
CAAA00593-FFEA
398.16
CAD
CAAA00593-FF
CAAA00593-FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.62 mm (0.024 in)
Erbium
MDL: MFCD00010987
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: ≈3.5g/25x25mm
Odor: Odorless
MDL No.: MFCD00010987
Formula:
Er MW: 167.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010987 CAS Number: 7440-52-0 EINECS: 231-160-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...