Dysprosium ≥99.8% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)

Supplier: Thermo Scientific Chemicals
Danger

00592-FF 00592-FI
CAAA00592-FFEA 290.72 CAD
CAAA00592-FF CAAA00592-FI
Dysprosium ≥99.8% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Dysprosium

MDL: MFCD00010982


Purity: 99.8% CAS No.: 7429-91-6 Physical Form: ≈3.3g/25x25mm, Packaged under argon Appearance: Gray Odor: Odorless UN No.: 3178 MDL No.: MFCD00010982

Formula: Dy
MW: 162.5
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3178
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR