Arsenic ≥99.999% (metals basis), lumps

Supplier: Thermo Scientific Chemicals
Danger

00590-14 00590-22
CAAA00590-14EA 270.18 CAD
CAAA00590-14 CAAA00590-22
Arsenic ≥99.999% (metals basis), lumps
Arsenic

MDL: MFCD00085309


Purity: 99.999% CAS No.: 7440-38-2 Physical Form: Ampouled under argon Appearance: Silver to black Odor: Odorless UN No.: 1558 MDL No.: MFCD00085309

Formula: As
Melting Pt: 817 °C
Density: 5.727 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II
Merck Index: 12,00832

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR