Tungsten ≥99.9% (metals basis), slug, Length: 6.4 mm (0.25 in), Ø 17.5 mm (0.689 in)

Supplier: Thermo Scientific Chemicals

00525-KG 00525-KI
CAAA00525-KGEA 206.98 CAD
CAAA00525-KG CAAA00525-KI
Tungsten ≥99.9% (metals basis), slug, Length: 6.4 mm (0.25 in), Ø 17.5 mm (0.689 in)
Tungsten

MDL: MFCD00011461


Purity: 99.9% CAS No.: 7440-33-7 Physical Form: ≈30g/slug MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR