Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), Thickness: 0.75 mm (0.03 in)
Supplier: Thermo Scientific Chemicals
00524-FF
00524-FI
CAAA00524-FFEA
167.48
CAD
CAAA00524-FF
CAAA00524-FI
Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), Thickness: 0.75 mm (0.03 in)
Hafnium
MDL: MFCD00011032
Purity: 99.5%
CAS No.: 7440-58-6
Physical Form: ≈6.1g/25x25mm
Odor: Odorless
MDL No.: MFCD00011032
Formula:
Hf Storage Temperature: Ambient |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...