Tungsten ≥99.95% (metals basis), wire, Ø 0.075 mm (0.003 in)

Supplier: Thermo Scientific Chemicals

00457-H4 00457-H9
CAAA00457-H4EA 145.83 CAD
CAAA00457-H4 CAAA00457-H9
Tungsten ≥99.95% (metals basis), wire, Ø 0.075 mm (0.003 in)
Tungsten

MDL: MFCD00011461


Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈0.09g/m MDL No.: MFCD00011461

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR