Antimony ≥99.9999% (metals basis), shot, Puratronic® 1.5-5 mm (0.06-0.2 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
00428-06
00428-14
00428-22
CAAA00428-06EA
69.52
CAD
CAAA00428-06
CAAA00428-14
CAAA00428-22
Antimony ≥99.9999% (metals basis), shot, Puratronic® 1.5-5 mm (0.06-0.2 in)
Antimony
MDL: MFCD00134030
Purity: 99.9999%
CAS No.: 7440-36-0
Odor: Odorless
MDL No.: MFCD00134030
Formula:
Sb Boiling Pt: 1637 °C (1013 hPa) Melting Pt: 630 °C Density: 6.684 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III Merck Index: 12,00733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...