Zirconium ≥99.95% (metals basis excluding Hf), wire (max. 0.25% Hf), Ø 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00419-G1 00419-G5
CAAA00419-G1EA 115.66 CAD
CAAA00419-G1 CAAA00419-G5
Zirconium ≥99.95% (metals basis excluding Hf), wire (max. 0.25% Hf), Ø 0.127 mm (0.005 in)
Zirconium

MDL: MFCD00011303


Purity: 99.95% CAS No.: 7440-67-7 Physical Form: ≈0.082g/m Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR