Zirconium ≥99.94% (metals basis excluding Hf), foil, thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

00417-FF 00417-FQ
CAAA00417-FFEA 130.82 CAD
CAAA00417-FF CAAA00417-FQ
Zirconium ≥99.94% (metals basis excluding Hf), foil, thickness 0.25 mm (0.01 in)
Zirconium

MDL: MFCD00011303


Purity: 99.94% CAS No.: 7440-67-7 Physical Form: ≈1g/25x25mm Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR