Zirconium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
00416-BW
00416-CB
CAAA00416-BWEA
158
CAD
CAAA00416-BW
CAAA00416-CB
Zirconium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Zirconium
MDL: MFCD00011303
Purity: 99.95%
CAS No.: 7440-67-7
Physical Form: ≈0.32g/m
Odor: Odorless
MDL No.: MFCD00011303
Formula:
Zr Storage Temperature: Ambient |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...