Ytterbium ≥99.9% (REO, rare earth oxide basis), ingot
Supplier: Thermo Scientific Chemicals
00407-06
00407-14
CAAA00407-06EA
149.63
CAD
CAAA00407-06
CAAA00407-14
Ytterbium ≥99.9% (REO, rare earth oxide basis), ingot
Ytterbium
MDL: MFCD00011286
Notes: May contain up to 0.15% Calcium
Purity: 99.9%
CAS No.: 7440-64-4
Physical Form: Packaged in air stable poly bottle
Odor: Odorless
MDL No.: MFCD00011286
Formula:
Yb Boiling Pt: 1427 °C (1013 hPa) Melting Pt: 824 °C Density: 6.977 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011286 CAS Number: 7440-64-4 EINECS: 231-173-2 Merck Index: 12,10239 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...