Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00401-FF 00401-FI
CAAA00401-FFEA 214.88 CAD
CAAA00401-FF CAAA00401-FI
Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)
Yttrium

MDL: MFCD00011468


Purity: 99.9% CAS No.: 7440-65-5 Physical Form: ≈0.36g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011468

Formula: Y
MW: 88.91
Boiling Pt: 2927
Melting Pt: 1525
Density: 4.472 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR