Yttrium ≥99.9% (REO, rare earth oxide basis), ingot

Supplier: Thermo Scientific Chemicals

00399-09 00399-18
CAAA00399-09EA 159.58 CAD
CAAA00399-09 CAAA00399-18
Yttrium ≥99.9% (REO, rare earth oxide basis), ingot
Yttrium

MDL: MFCD00011468 Contains less than 1% tantalum


Purity: 99.9% CAS No.: 7440-65-5 Molecular Formula: Contains less than 1% tantalum Physical Form: Packaged in air stable poly bottle Odor: Odorless

Formula: Y
Boiling Pt: 2927 °C (1013 hPa)
Melting Pt: 1525 °C
Density: 4.472 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR